Learn More
Agilent BioTek Spare syringe pump tubing
Product Code. 12516586 Shop All Agilent BioTek Products
Click to view available options
For Use With (Equipment):
MicroFill
Unit Size:
Each
Product Code. 12516586
Brand: Agilent BioTek 7140514
Only null left
Add to basket
Add to basket
Spare syringe pump dispense tubing and check valve kit (71405140)
Organic solvent compatible (non-autoclavable, but compatible with the autoclavable syringe module hardware)
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Not Autoclavable
Description Spare syringe pump dispense tubing and check valve kit
For Use With (Equipment) MicroFill