missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Supplemented Brucella Broth for Anaerobes

Product Code. 16082272
Change view
Click to view available options
Quantity:
10 mL
Unit Size:
10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16082272 10 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16082272 Supplier Thermo Scientific™ Supplier No. YT3450
  • ONLINE EXCLUSIVE PRICE
    £98.75
    £64.48 / 10mL
    Save £34.27
    34% Off
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Perform susceptibility testing with a full range of supplies, including this broth, to enhance your Thermo Scientific™ Sensititre™ test program.

Perform high-quality susceptibility testing with a complete range of supplies, including the Thermo Scientific™ Supplemented Brucella Broth for Anaerobes, to complement and enhance your Thermo Scientific™ Sensititre™ test program.

Not all products are available for sale in all territories. Please inquire.

VersaTREK™ and Sensititre products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY

Specifications

Description Supplemented Brucella Broth for Anaerobes
Product Type Microbiology Media
Target Organism Class Anaerobes
Test Type Microbial Susceptibility
No. per Pack 10
Packaging Type 10 mL/Bottle
Quantity 10 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.