missing translation for 'onlineSavingsMsg'
Learn More

Suplatast (Tosilate), MedChemExpress

Product Code. 16427226
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16427226 100 mg 100mg
16421826 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16427226 Supplier Medchem Express Supplier No. HY17002

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Suplatast Tosilate (IPD 1151T) is an orally active Th2 cytokine inhibitor which can inhibit both IL-4 and IL-5 production from Th2 cells and suppress IgE synthesis. Suplatast Tosilate is an anti-allergic agent. Suplatast Tosilate has antiasthmatic, anti-inflammatory and antifibrotic activity.

Suplatast Tosilate (IPD 1151T) is an orally active Th2 cytokine inhibitor which can inhibit both IL-4 and IL-5 production from Th2 cells and suppress IgE synthesis. Suplatast Tosilate is an anti-allergic agent. Suplatast Tosilate has antiasthmatic, anti-inflammatory and antifibrotic activity.

Specifications

Chemical Name or Material Suplatast (Tosilate)
CAS 94055-76-2
Color White
Purity Grade Notes Research
Molecular Formula C23H33NO7S2
Quantity 100 mg
Synonym IPD 1151T
Solubility Information DMSO : ≥ 33 mg/mL (66.05 mM)
SMILES C[S+](C)CCC(NC1=CC=C(C=C1)OCC(COCC)O)=O.CC2=CC=C(C=C2)S(=O)([O-])=O
Molecular Weight (g/mol) 499.64
Formula Weight 499.64
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.