missing translation for 'onlineSavingsMsg'
Learn More

Labcon™ SuperSlik™ 250 μL Low Retention Pipette Tips

Product Code. 17517876
Click to view available options
Sterility:
Non-sterile
Sterile
Format:
Bagged (10,000)
Rack (9600)
Refill (4800)
Refill (9600)
Unit Size:
Pack of 1000
Pack of 480
Pack of 960
This item is not returnable. View return policy

Product Code. 17517876

Brand: Labcon™ 11669650069

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

These low retention pipette tips with UltraFine™ points come in resealable bags or racks.

SuperSlik™ Low Retention tips UltraFine™ Point Fits Popular 250 uL Pipettes Protease, Human DNA, ATP, RNase/DNase and Endotoxin Free (non-pyrogenic) Polypropylene USP Class VI resin Autoclavable 52.1 mm H. Made in USA

Specifications

Volume (Metric) 250 μL
Filtered Non-filtered
Sterility Sterile
Color Clear
Autoclavable Autoclavable
Length (English) 2 in.
Length (Metric) 52.1 mm
Certifications/Compliance ISO 9001:2015, FDA CFR21 GMP, USP Class VI
Clean Claims Protease, Human DNA, ATP, RNase/DNase, Endotoxin-free
For Use With (Application) Micropipetting
Sterilization Method E-beam
Format Rack (9600)
Quantity 96 Tips/Rack, 10 Racks/Pk., 10 Pk./Cs.
Green Features Manufactured with 93% Renewable Energy
Disposable Single-use
Graduated No
Material Polypropylene Resin
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.