missing translation for 'onlineSavingsMsg'
Learn More

Labcon™ SuperSlik™ 10 μL Extra Long Low Retention Pipette Tips

Product Code. 17567866
Click to view available options
Sterility:
Non-sterile
Sterile
Format:
Bagged
Rack, 96
Unit Size:
Pack of 1000
Pack of 960
This item is not returnable. View return policy

Product Code. 17567866

Brand: Labcon™ 11659650069

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

These graduated, extra long pipette tips with UltraFine™ Points come in resealable bags or racks.

  • Graduated at 2 μL and 10 μL
  • Fits standard 10 μL pipettes
  • If sterile, have undergone Validated Sterilization (ISO 11137) for use in Research and Medical labs
  • Polypropylene USP Class VI Resin
  • Made in USA

Specifications

Volume (Metric) 10 μL
Filtered Non-filtered
Sterility Sterile
Color Clear
Rack Color Purple
Autoclavable Autoclavable
Length (English) 2 in.
Length (Metric) 47.6 mm
Certifications/Compliance ISO 9001:2015, FDA CFR21 GMP, USP Class VI
Clean Claims Protease, Human DNA, ATP, RNase/DNase, Endotoxin-free
For Use With (Application) Micropipetting
Sterilization Method E-beam
Format Rack, 96
Tip Style Extended Length
Quantity 96 Tips/Rack, 10 Racks/Pk., 10 Pk./Cs.
Disposable Single-use
Graduated Yes
Material Polypropylene Resin
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.