Learn More
Thermo Scientific™ SuperSignal™ West Pico Chemiluminescent Substrate
Shop All Thermo Scientific ProductsDescription
Thermo Scientific™ SuperSignal West Pico Chemiluminescent Substrate is an enhanced chemiluminescent (ECL) substrate for horseradish peroxidase (HRP) enzyme that provides low picogram detection of proteins in Western blot applications.
Highlights:
ECL substrate – an enhanced chemiluminescent substrate for detection of horseradish peroxidase (HRP) activity from antibodies and other Western blot probes
Economy – costs less per milliliter than other similarly sensitive chemiluminescent substrates
Picogram sensitivity – highly sensitive for the rapid development of a wide range of protein levels
Stable light output – long-duration signal makes it possible to make multiple exposures to film
High intensity – signal is twice as strong as output from other luminol-based systems
Excellent stability – 24-hour working solution stability; 1-year kit stability at room temperature
Saves antibody – requires much less (more dilute) primary and secondary antibodies per blot
Specifications
Specifications
| Description | SuperSignal West Pico Chemiluminescent Substrate |
| Detection Method | Chemiluminescence |
| Quantity | 50 mL Kit |
| Conjugate | HRP |
| Sufficient For | 6 mini-gel blots/500 cm2 membrane |
| Kit Contents | Luminol/Enhancer, 25mL, Stable Peroxide Buffer, 25mL |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.