missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ SuperSignal™ West Pico Chemiluminescent Substrate

Product Code. 11859280 Shop All Thermo Scientific Products
Click to view available options
Quantity:
1 L Kit
Unit Size:
1 L
This item is not returnable. View return policy

Product Code. 11859280

Brand: Thermo Scientific™ 34078

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Enhanced chemiluminescent (ECL) HRP substrate that provides low-picogram-level detection by Western blot analysis. These products will be discontinued on September 30th, 2017. Please switch to the longer-lasting, more-sensitive SuperSignal West Pico PLUS.

Thermo Scientific™ SuperSignal West Pico Chemiluminescent Substrate is an enhanced chemiluminescent (ECL) substrate for horseradish peroxidase (HRP) enzyme that provides low picogram detection of proteins in Western blot applications.

SuperSignal West Pico Substrate emits light for longer and chemiluminescent signal at more than twice the intensity of other entry-level luminol-based horseradish peroxidase detection systems while also maintaining sensitivity and low background. In addition, the greater signal intensity is achieved using less primary and secondary antibody than typical ECL substrates. All of this for about 30% less cost than comparable products.

Highlights:

  • ECL substrate – an enhanced chemiluminescent substrate for detection of horseradish peroxidase (HRP) activity from antibodies and other Western blot probes

  • Economy – costs less per milliliter than other similarly sensitive chemiluminescent substrates

  • Picogram sensitivity – highly sensitive for the rapid development of a wide range of protein levels

  • Stable light output – long-duration signal makes it possible to make multiple exposures to film

  • High intensity – signal is twice as strong as output from other luminol-based systems

  • Excellent stability – 24-hour working solution stability; 1-year kit stability at room temperature

  • Saves antibody – requires much less (more dilute) primary and secondary antibodies per blot

TRUSTED_SUSTAINABILITY

Specifications

Description SuperSignal West Pico Chemiluminescent Substrate
Detection Method Chemiluminescence
Quantity 1 L Kit
Conjugate HRP
Sufficient For 120 mini-gel blots; 10,000cm2 of membrane
Kit Contents Stable Peroxide Buffer, 500mL, Luminol/Enhancer, 500mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.