Learn More
Thermo Scientific™ SuperSignal™ West Pico Chemiluminescent Substrate
Shop All Thermo Scientific ProductsDescription
Thermo Scientific™ SuperSignal West Pico Chemiluminescent Substrate is an enhanced chemiluminescent (ECL) substrate for horseradish peroxidase (HRP) enzyme that provides low picogram detection of proteins in Western blot applications.
Highlights:
ECL substrate – an enhanced chemiluminescent substrate for detection of horseradish peroxidase (HRP) activity from antibodies and other Western blot probes
Economy – costs less per milliliter than other similarly sensitive chemiluminescent substrates
Picogram sensitivity – highly sensitive for the rapid development of a wide range of protein levels
Stable light output – long-duration signal makes it possible to make multiple exposures to film
High intensity – signal is twice as strong as output from other luminol-based systems
Excellent stability – 24-hour working solution stability; 1-year kit stability at room temperature
Saves antibody – requires much less (more dilute) primary and secondary antibodies per blot
Specifications
Specifications
| Description | SuperSignal West Pico Chemiluminescent Substrate |
| Detection Method | Chemiluminescence |
| Quantity | 100 mL Kit |
| Conjugate | HRP |
| Sufficient For | 12 mini-gel blots/1000 cm2 membrane |
| Kit Contents | Luminol/Enhancer, 50mL, Stable Peroxide Buffer, 50mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.