missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SuperScript V Reverse Transcriptase

Product Code. 18951133
Change view
Click to view available options
Quantity:
10,000 Units
2000 Units
40,000 Units
Unit Size:
10 reactions
200 reactions
50 reactions
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18951133 40,000 Units 200 reactions
18931133 2000 Units 10 reactions
18941133 10,000 Units 50 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 18951133 Supplier Invitrogen™ Supplier No. 19090200
 Restricted Item
Estimated Shipment: 16-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

Invitrogen™ SuperScript™ V Reverse Transcriptase (RT) is a next-generation MMLV reverse transcriptase designed for fast, reliable first-strand cDNA synthesis.

Invitrogen™ SuperScript™ V Reverse Transcriptase (RT) is a next-generation MMLV reverse transcriptase designed for fast, reliable first-strand cDNA synthesis. Building on the advanced performance of the SuperScript™ IV reverse transcriptase, SuperScript V RT combines high sensitivity, thermostability, and processivity with improved resistance to oxidation and greater workflow flexibility.

SuperScript V RT supports reliable cDNA synthesis from low-input RNA as well as highly structured, GC-rich, degraded, and inhibitor-containing samples.

Features of SuperScript V Reverse Transcriptase include:

  • Broad RNA input range—supports cDNA synthesis from 100 fg to 5 μg of RNA
  • Robust performance with challenging RNA—reaction temperatures up to 65°C help transcribe highly structured and GC-rich RNA
  • Fast cDNA synthesis—complete first-strand cDNA synthesis in as little as 1–10 minutes
  • Improved resistance to oxidation— enhanced reagent stability supports ambient shipping and room-temperature storage for up to 6 months
  • Flexible workflow—room-temperature reaction setup and stability of assembled reactions for up to 24 hours

SuperScript V RT is well suited for RT-PCR, RT-qPCR, gene expression analysis, and other applications requiring high-quality first-strand cDNA synthesis. Its enhanced reagent and assembled-reaction stability provides additional flexibility for routine, high-throughput, and automated workflows.

A DTT-free, animal origin–free formulation combined with reduced cold chain requirements further simplifies reagent handling and supports more sustainable workflows.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 200 U/μL
Content And Storage • SuperScript V Reverse Transcriptase (200 μL)
• 5X SuperScript V Reaction Buffer (2 x 1 mL)

Store all contents at:
• -20°C for long-term storage until expiry date
• +5°C for up to 12 months
• Room temperature for up to 6 months
Detection Method qPCR, NGS, PCR
Form Liquid
Format Stand-alone Enzyme
GC-Rich PCR Performance High
PCR Method 2-step RT-qPCR, RT-PCR
Reaction Speed 10 min.
Technique Reverse Transcription
Label or Dye Not Applicable
Optimal Reaction Temperature 50°C
Product Type Reverse Transcription Reagent
Quantity 40,000 Units
Reverse Transcriptase SuperScript V
Ribonuclease H Activity Reduced
Sensitivity 0.1 pg total RNA
Shipping Condition Dry Ice, Wet Ice, Ambient
For Use With (Application) cDNA Synthesis, RT-PCR, Real Time PCR (qPCR), Sequencing, Next-Generation Sequencing (NGS)
Thermostability Up to 65°
No. of Reactions 200 Reactions
Reaction Format Separate components
Reagent Type Reverse Transcription
Size (Final Product) Up to 20 kb
Starting Material RNA
Inhibitor Tolerance (Simple) High
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.