missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SuperScript™ III First-Strand Synthesis SuperMix for qRT-PCR

Product Code. p-4926553
Change view
Click to view available options
No. of Reactions:
250 Reactions
50 Reactions
Unit Size:
250 reactions
50 reactions
Product Code. No. of Reactions unitSize
10246902 50 Reactions 50 reactions
10338102 250 Reactions 250 reactions
2 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10246902

Brand: Invitrogen™ 11752050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

SuperScript III First-Strand Synthesis SuperMix for qRT-PCR provides the high-temperature capability of SuperScript III Reverse Transcriptase in an optimized SuperMix format for the synthesis of first-strand cDNA for use in real-time quantitative RT-PCR (qRT-PCR).

SuperScript III First-Strand Synthesis SuperMix for qRT-PCR provides the high-temperature capability of SuperScript III Reverse Transcriptase in an optimized SuperMix format for the synthesis of first-strand cDNA for use in real-time quantitative RT-PCR (qRT-PCR). The simple, time-saving reaction set-up uses just two tubes: a 2X reaction mix and an enzyme mix.

Enzyme mix
SuperScript III Reverse Transcriptase, included in the RT enzyme mix, is a version of M-MLV RT that has been engineered to reduce RNase H activity and provide increased thermal stability. The enzyme can be used to synthesize cDNA at a temperature range of 42–60°C, providing increased specificity, higher yields of cDNA, and more full-length product than other reverse transcriptases. Because SuperScript III RT is not significantly inhibited by ribosomal and transfer RNA, it can be used to synthesize cDNA from total RNA. RNaseOUT Recombinant Ribonuclease Inhibitor, also included in the enzyme mix, is an RNase inhibitor protein that safeguards against the degradation of target RNA due to ribonuclease contamination of the RNA preparation.

Reaction mix
The 2X RT reaction mix includes oligo(dT)20, random hexamers, MgCl2, and dNTPs in a buffer formulation that has been optimized for qRT-PCR. E. coli RNase H is provided as a separate tube in the kit to remove the RNA template from the cDNA:RNA hybrid molecule after first-strand synthesis. This has been shown to increase sensitivity in qRT-PCR.

Using SuperScript III First-Strand Synthesis SuperMix
This SuperMix formulation can be used to quantify fewer than 10 copies of a target gene in qRT-PCR, with a broad dynamic range that supports accurate quantification of high-copy mRNA from up to 1 μg of total RNA. Reagents are provided for 50 or 250 RT reactions of 20 μL each.

Order Info

Shipping Condition: Dry ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage

• 2X RT Reaction Mix (500 μL)
• RT Enzyme Mix (100 μL)
E. coli RNase (50 μL)

Store at –20°C.

Detection Method qPCR
Format Super Mix
GC-Rich PCR Performance High
Reaction Speed 30 min.
Technique Reverse Transcription
Optimal Reaction Temperature 50°C
Quantity 50 rxns
Reverse Transcriptase SuperScript III
Shipping Condition Dry Ice
For Use With (Application) Real Time PCR (qPCR)
Final Product Type First-Strand cDNA
No. of Reactions 50 Reactions
Reaction Format Master Mix
Reagent Type Reverse Transcription
Starting Material RNA
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.