missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SuperScript™ III CellsDirect™ cDNA Synthesis Kit

Product Code. 10442482
Change view
Click to view available options
No. of Reactions:
100 Reactions
25 Reactions
Unit Size:
1 set
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Reactions unitSize
10442482 25 Reactions 1 set
10694993 100 Reactions 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10442482 Supplier Invitrogen™ Supplier No. 18080200

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

The SuperScript III CellsDirect cDNA Synthesis Kit is optimized for synthesis of first-strand cDNA directly from a mammalian cell lysate without first isolating the RNA.

The SuperScript III CellsDirect cDNA Synthesis Kit is optimized for synthesis of first-strand cDNA directly from a mammalian cell lysate without first isolating the RNA. Lysis and reverse transcription are performed in the same tube, and the resulting first-strand cDNA is ready to use in cloning and PCR.

Features of the SuperScript III CellsDirect Synthesis Kit include:
• Compatible with a wide range of mammalian cell types grown under different treatment conditions
• Single-tube format minimizes reagent loss, sample loss, and handling time
• Total lysate volume is used in first-strand cDNA synthesis reaction, providing greater yields with a limited number of cells and allowing for detection of rare transcripts
• SuperScript III Reverse Transcriptase, with reduced RNase H activity and higher thermal stability, produces high yields of cDNA in the first-strand synthesis reaction, for greater sensitivity and enhanced detection of rare transcripts
• Generates high-quality cDNA for use in a variety of applications, including cloning and PCR
• Simple protocol takes less than two hours

How it works
In traditional RT-PCR, RNA is first isolated from cells in a time-consuming procedure that can lead to a loss of material. Using the SuperScript III CellsDirect cDNA Synthesis Kit, the cells are lysed and the cDNA is generated from the lysate in a single tube with minimal handling and no sample loss. DNase I is added to eliminate genomic DNA prior to first-strand synthesis. This kit has been optimized for small cell samples, ranging from 10,000 cells down to a single cell (as measured by serial dilution). The use of SuperScript III Reverse Transcriptase ensures high specificity and high yields of cDNA from small amounts of starting material (as little as 10 pg total RNA). After synthesis, the first-strand cDNA can be amplified with specific primers by PCR without intermediate organic extractions or ethanol precipitations.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage

• SuperScript III RT
• RNaseOUT Ribonuclease Inhibitor
• Oligo(dT)20 primers
• DNase I
• DNase I buffer
• EDTA
• dNTP mix
• 5X First-strand buffer
• DTT (0.1 M)
• RNase H
• Control RNA & primers
• Resuspension buffer

Store at –20°C.
Guaranteed stable for 6 months when properly stored.

Format Kit
GC-Rich PCR Performance High
Reaction Speed 50 min.
Technique Reverse Transcription
Optimal Reaction Temperature 50°C
Quantity 25 rxns
Reverse Transcriptase SuperScript III
Ribonuclease H Activity Reduced
For Use With (Application) Real Time PCR (qPCR)
Final Product Type First-Strand cDNA
No. of Reactions 25 Reactions
Reaction Format Separate components
Reagent Type Reverse Transcription
Size (Final Product) Up to 4.5 kb
Starting Material Cells
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.