missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Superpack Pipette Tips
Shop All MP Biomedicals Products
Click to view available options
Sterility:
Non-sterile
Sterile
Quantity:
10 x 96/Box
1000/Box
Unit Size:
Pack of 1000
Pack of 960
Description
Superior pipette tips
- Autoclavable
- Recyclable
- Very low coefficient of friction
- Naturally hydrophobic
- Metal-free
Specifications
Specifications
| Sterility | Non-sterile |
| Color | Natural |
| Autoclavable | Autoclavable |
| Quantity | 10 x 96/Box |
| Product Line | Superpack |
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction