missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Superoxide Dismutase (Bovine)

Product Code. 16131790
Click to view available options
Quantity:
2 mg
Unit Size:
2mg
This item is not returnable. View return policy

Product Code. 16131790

Brand: Abnova™ P5252.2mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used for Enzyme

#REF!

Specifications

For Use With (Application) Enzyme Activity
Molecular Weight (g/mol) 32.5kDa
Name Superoxide Dismutase (Bovine)
Preparation Method Native protein chromatographically purified from Bovine Erythrocytes essentially as described by McCord and Fridovich, J. Biol. Chem., 244, 6049 (1969)
Quantity 2 mg
Storage Requirements Store at 4°C.
Regulatory Status RUO
Common Name SUPEROXIDE
Biological Activity ≥1,400 units per mg dry weight. One Unit inhibits by 50% the maximum reduction of nitro blue tetrazolium under the specified conditions.
Species Bovine
Protein Tag None
Expression System Native protein chromatographically purified from Bovine Erythrocytes essentially as described by McCord and Fridovich, J. Biol. Chem., 244, 6049 (1969)
Form Lyophilized
Show More Show Less
compliance-icons
Product Identifier
  • 2MG Superoxide Dismutase
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P233-Keep container tightly closed.
  • P264-Wash hands thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P404-Store in a closed container.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : sodium phosphate dibasic, potassium phosphate monobasic
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.