missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Superose™ Media Prep Grade Lab Packs

Product Code. 11544915 Shop All Cytiva Products
Change view
Click to view available options
Product Type:
Superose 12 Prep Grade
Superose 6 Prep Grade
Max. Pressure:
100 psi
58 psi
Unit Size:
125mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Max. Pressure Unit Size
11544915 Superose 6 Prep Grade 58 psi 125mL
11514925 Superose 12 Prep Grade 100 psi 125mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11544915 Supplier Cytiva Supplier No. 17048901
Powering Proteomics
Use promo code 28003
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

High performance gel filtration media for scaleup to preparative purifications of biomolecules

  • Permit high recovery of a wide variety of biomolecules—proteins, peptides, polynucleotides, etc.
  • Supplied as bulk filtration media, for preparative separations
  • Intended as a complement to Superose PC prepacked columns; can be efficiently packed under medium pressure
  • Superose 6 Prep Grade intended for biomolecules of a wide fractionation range; Superose 12, for higher-molecular weight proteins

Order Info

Supplied in 125mL containers

TRUSTED_SUSTAINABILITY

Specifications

Quantity Each
Flow Rate 40 cm/hr.
Particle Size 30 to 40 μm
For Use With (Equipment) AKTA™ design pump or system
Bed Volume ≈100 mL
Product Type Superose 6 Prep Grade
For Use With (Application) Separations of Proteins and other biomolecules according to size
Max. Pressure 58 psi
pH Range 3 to 12
Support Type Highly Cross-linked Agarose
Sample Type Proteins, Peptides, Polynucleotides, Wide Variety of Biomolecules
Content And Storage 4 to 30°C, 20% Ethanol
Show More Show Less
compliance-icons
Product Identifier
  • Superose 6 prep grade, 125 ml
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: ethanol
  • UNKNOWN ACUTE-16.5 per cent of the mixture consists of component(s) of unknown acute toxicity.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.