Learn More
Epredia™ SuperFrost™ Microscope Slides, Ground 90°
The printed Epredia™ SuperFrost tab is impervious to virtually all common laboratory chemicals and reagents. A wide range of colors (7 standard colors) allows several examination specimens to be labeled and archived simply and clearly.
Brand: Epredia™ AG00008132E01FST20
Code : 03
Additional Details : Weight : 0.22500kg
Description
- Time-consuming labeling with paper labels is no longer required
- The writing area is resistant to all common solvents
- SuperFrost slides may be sorted by color e.g. by patient, urgency, stain or examination method
- Simple recognition of specified characteristics such as year of examination or department, use etc.
Specifications
90°, Ground | |
76 mm | |
Blue | |
Uncoated | |
Yes |
1 mm | |
26 mm | |
ISO8037/I | |
50/Box, 2000 in outer carton | |
76 x 26 mm |