Learn More
Epredia™ SuperFrost™ Plus Slides
Product Code. 12608106 Shop All Epredia Products
Change view
Click to view available options
:
White; 144/cs
Product Code.
12608106 White; 144/cs
1 options
Product Code. 12608106 Supplier Epredia™ Supplier No. 6776214CS
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Epredia™ Superfrost™ Plus Slides are positively charged for improved cellular adhesion.
  • Electrostatically attract tissue sections and cytology preparations, binding them to the slide
  • Eliminate the need for special adhesives or protein coatings
  • No background staining
  • Available in a variety of colors
TRUSTED_SUSTAINABILITY
Specifications
Frosted Yes
Type Microscope Slide
Color White
Quantity 1440/Cs.