Learn More
GE Healthcare SuperFrac™ Tube Racks
Product Code. 12480970 Shop All Cytiva Products
Click to view available options
Holds:
Tubes (12 to 13 mm)
Unit Size:
Pack of 12
Product Code. 12480970
Brand: GE Healthcare 18101020
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Product Type Tube Rack
Holds Tubes (12 to 13 mm)
Positions 26
Quantity 12 Pack
Diameter (Metric) Opening 13 mm