missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Superdex™ 75 Prep Grade

Product Code. 11580804 Shop All Cytiva Products
Click to view available options
Quantity:
150 mL
Unit Size:
150mL
This item is not returnable. View return policy

Product Code. 11580804

Brand: Cytiva 17104401

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Offers high resolution fractionation for separation, polishing and product formulation of recombinant proteins

  • Gel filtration medium
  • For purifying proteins of fractionation range: 3000 to 70000Da
  • Average particle size: 34μm
  • Matrix: composite of cross-linked agarose and dextran
  • pH stability working range
  • Available in 150mL bottle

TRUSTED_SUSTAINABILITY

Specifications

Quantity 150 mL
Flow Rate 10 to 50 cm/hr.
Particle Size 24 to 44 μm
Product Type Superdex Media
Format Solid Spherical Bead
For Use With (Application) Separation, polishing and product formulation of recombinant Proteins
Max. Pressure 70psi
pH Range 3 to 12
Support Type Composite of cross-linked Agarose and Dextran
Sample Type Recombinant Protein
Content And Storage 4 to 30°C, 0.2 M sodium acetate in 20% Ethanol
Show More Show Less
compliance-icons
Product Identifier
  • Superdex 75 prep grade, 150 ml
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : ethanol
  • UNKNOWN ACUTE-18 per cent of the mixture consists of component(s) of unknown acute toxicity.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.