Learn More
Specac™ Supercritical Fluids Diamond ATR Top Plate
Product Code. 15230118
Encompass_Preferred
Click to view available options
Type:
Top Plate
Product Code. 15230118
Brand: Specac™ GS10585
This item requires a price update prior to being ordered.
Request a Price for this item here.
The Supercritical Fluids Analyzer Top-Plate option for the Golden Gate Diamond ATR GS10500 is designed for operation at extreme temperatures and pressures. The Diamond ATR element is high temperature metal bonded into a tungsten carbide disk, avoiding the use of adhesives or resins that may be dissolved or attacked by the chemical aggressiveness of supercritical fluids. Enables pressures up to 6000 psi and temperatures up to 300°C.
Specifications
Type Top Plate
Description Enables pressures up to 6000 psi and temperatures up to 300°C