missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Super AquaBlue ELISA Substrate
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
Description
- Highly sensitive and extremely stable (18 months at room temperature or 24 months at 4°C).
- Engineered for convenience as a single component (no additional H2O2 or other components are necessary)
- Suitable for endpoint reactions, but is also ideal for kinetic assays; stopping the reaction with acid stop solution does not change the color or absorption properties of the blue-green end product
Especificaciones
Especificaciones
| Product Type | ELISA Substrate |
| Quantity | 500 mL |
| Detection Method | Colorimetric |
| Substrate Properties | Chemical Substrate |
| Substrate Type | HRP (Horseradish Peroxidase) |
For Research Use Only. Not for use in diagnostic procedures.
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido