Learn More
Cellpath Supa Mega Tissue Processing Cassettes
Product Code. p-4908708
Change view
Click to view available options
Color:
White
Unit Size:
Pack of 100
Product Code. Color unitSize
30226233 White Pack of 100
1 options
Product Code. 30226233 Supplier Cellpath Supplier No. EAG010202A
Add to basket
Edge
Add to basket
Widely adopted in research and toxicology laboratories. CellPath Supa Mega Tissue Processing Cassettes are ideal for embedding and processing larger tissue samples of prostrate, brain, breast and eye.
  • Plastic hinged lid
  • Rigid, durable
  • Excellent writing surfaces
  • Internal chamber is three times depth of the Supa Mega Slim and Supa Mega Mother Ship
  • Available in a range of colors
Specifications
Quantity 100/Pk.
Color White