missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SUOX Recombinant Rabbit Monoclonal Antibody (24GB1790)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557746
This item is not returnable.
View return policy
Description
SUOX Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
Sulfite oxidase is a homodimeric protein localized to the intermembrane space of mitochondria. Each subunit contains a heme domain and a molybdopterin-binding domain. The enzyme catalyzes the oxidation of sulfite to sulfate, the final reaction in the oxidative degradation of the sulfur amino acids cysteine and methionine. Sulfite oxidase deficiency results in neurological abnormalities which are often fatal at an early age. Alternative splicing results in multiple transcript variants encoding identical proteins.
Specifications
| SUOX | |
| Recombinant Monoclonal | |
| Unconjugated | |
| SUOX | |
| SO; sulfite oxidase; sulfite oxidase, mitochondrial; Suox | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 6821 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB1790 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P51687 | |
| SUOX | |
| A synthesized peptide derived from human SUOX (aa 150-190). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction