missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SUOX Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15075-1-AP-150UL
This item is not returnable.
View return policy
Description
Sulfite oxidase is a homodimeric protein localized to the intermembrane space of mitochondria. Each subunit contains a heme domain and a molybdopterin-binding domain. The enzyme catalyzes the oxidation of sulfite to sulfate, the final reaction in the oxidative degradation of the sulfur amino acids cysteine and methionine. Sulfite oxidase deficiency results in neurological abnormalities which are often fatal at an early age. Alternative splicing results in multiple transcript variants encoding identical proteins.Specifications
| SUOX | |
| Polyclonal | |
| Unconjugated | |
| SUOX | |
| sulfite oxidase, Sulfite oxidase, mitochondrial, SUOX | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 211389, 6821, 81805 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation | |
| 0.75 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P51687, Q07116, Q8R086 | |
| Suox | |
| SUOX Fusion Protein Ag7146 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction