missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SUOX Monoclonal Antibody (OTI1A12)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA526763
This item is not returnable.
View return policy
Description
SUOX Monoclonal Antibody for Western Blot, IHC (P)
Sulfite oxidase is a homodimeric protein localized to the intermembrane space of mitochondria. Each subunit contains a heme domain and a molybdopterin-binding domain. The enzyme catalyzes the oxidation of sulfite to sulfate, the final reaction in the oxidative degradation of the sulfur amino acids cysteine and methionine. Sulfite oxidase deficiency results in neurological abnormalities which are often fatal at an early age. Alternative splicing results in multiple transcript variants encoding identical proteins.
Specifications
| SUOX | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P51687 | |
| SUOX | |
| Human recombinant protein fragment corresponding to amino acids 320-545 of SUOX produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI1A12 | |
| Unconjugated | |
| SUOX | |
| SO; sulfite oxidase; sulfite oxidase, mitochondrial; Suox | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 6821 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction