missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sunitinib, MedChemExpress
Click to view available options
Quantity:
100 mg
200 mg
500 mg
Unit Size:
100mg
200mg
500mg
Description
Sunitinib (SU 11248) is a multi-targeted receptor tyrosine kinase inhibitor with IC50s of 80 nM and 2 nM for VEGFR2 and PDGFRβ, respectively. Sunitinib, an ATP-competitive inhibitor, effectively inhibits autophosphorylation of Ire1α by inhibiting autophosphorylation and consequent RNase activation.
Specifications
Specifications
| Chemical Name or Material | Sunitinib |
| CAS | 557795-19-4 |
| Color | Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C22H27FN4O2 |
| Quantity | 200 mg |
| Synonym | SU 11248 |
| Solubility Information | DMSO : 20.83 mg/mL (52.27 mM; ultrasonic and warming and heat to 80°C) |
| SMILES | O=C(NCCN(CC)CC)C1=C(NC(/C=C2C(NC3=C\2C=C(C=C3)F)=O)=C1C)C |
| Molecular Weight (g/mol) | 398.47 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction