missing translation for 'onlineSavingsMsg'
Learn More

Sunifiram, Mikromol™

Product Code. 30051906
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30051906 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30051906 Supplier Mikromol Supplier No. MM3274.00

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Sunifiram
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sunifiram
CAS 314728-85-3
Analyte or Component Names Sunifiram
Shipping Condition Room Temperature
Molecular Formula C14 H18 N2 O2
Certifications/Compliance ISO 17025
Synonym 1-Propanone, 1-(4-benzoyl-1-piperazinyl)-, Piperazine, 1-benzoyl-4-(1-oxopropyl)- (9CI), DM 235, Sunifiram
InChI Formula InChI=1S/C14H18N2O2/c1-2-13(17)15-8-10-16(11-9-15)14(18)12-6-4-3-5-7-12/h3-7H,2,8-11H2,1H3
SMILES CCC(=O)N1CCN(CC1)C(=O)c2ccccc2
IUPAC Name 1-(4-benzoylpiperazin-1-yl)propan-1-one
Molecular Weight (g/mol) 246.3
Quantity 100 mg
Formula Weight 246.1368 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.