Learn More
Thermo Scientific™ SUN-SRi™ 13-425 Screw Thread Caps
Product Code. 31920938 Shop All Thermo Scientific Products
Change view
Click to view available options
Color:
Black
Product Type:
Solid Top Cap
Material:
Phenolic
Unit Size:
Pack of 100
Product Code. Color Product Type Material Unit Size
31920938 Black Solid Top Cap Phenolic Pack of 100
1 options
Product Code. 31920938 Supplier Thermo Scientific™ Supplier No. 200 568
Estimated Shipment: 23-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Thermo Scientific caps and vials are available in a wide variety of sizes and configurations to fit the needs of many applications.
TRUSTED_SUSTAINABILITY
Specifications
Material (Liner) PTFE/Rubber
Color Black
Material Phenolic
Closure Size 13-425
Product Type Solid Top Cap