missing translation for 'onlineSavingsMsg'
Learn More

SUMO3 Antibody (401513R) [DyLight 680], Novus Biologicals™

Product Code. 30737756 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30737756 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30737756 Supplier Novus Biologicals Supplier No. FAB2959RY

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

SUMO3 Monoclonal antibody specifically detects SUMO3 in Human samples. It is validated for Immunocytochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen SUMO3
Applications Immunocytochemistry
Classification Monoclonal
Clone 401513R
Conjugate DyLight 680
Formulation 50mM Sodium Borate
Gene Alias SMT3 (suppressor of mif two 3, yeast) homolog 1, SMT3 homolog 1, SMT3 suppressor of mif two 3 homolog 1, SMT3 suppressor of mif two 3 homolog 3 (S. cerevisiae), SMT3 suppressor of mif two 3 homolog 3 (yeast), SMT3A, Smt3B, SMT3H1small ubiquitin-related modifier 3, SUMO-2, SUMO-3, ubiquitin-like protein SMT3A, Ubiquitin-like protein SMT3B
Host Species Rat
Immunogen E. coli-derived recombinant human SUMO3, Met1-Phe103, Accession # P55854
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 6612
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.