missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
SUMO2/3 Monoclonal antibody specifically detects SUMO2/3 in Human samples. It is validated for Flow Cytometry
Specifications
Specifications
| Antigen | SUMO2/3 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 10F1 |
| Conjugate | mFluor Violet 450 SE |
| Dilution | Flow Cytometry |
| Formulation | 50mM Sodium Borate |
| Gene Alias | HSMT3, small ubiquitin-like modifier 2, SMT3A, SMT3B, SMT3H2, SUMO2, SUMO3 |
| Host Species | Mouse |
| Immunogen | Recombinant human SUMO2/3 (1-93aa) purified from E.coli |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?