missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SUMO1 Rabbit anti-Human, Clone: 3, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90170-50UL
This item is not returnable.
View return policy
Description
SUMO1 Monoclonal specifically detects SUMO1 in Human samples. It is validated for ELISA.
Specifications
| SUMO1 | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 3 | |
| ELISA 1:5000-1:10000 | |
| DAP1, GAP modifying protein 1, GAP-modifying protein 1, GMP1SMT3CSMT3H3OFC10UBL1PIC1, SENP2, sentrin, small ubiquitin-related modifier 1, SMT3, SMT3 homolog 3, SMT3 suppressor of mif two 3 homolog 1 (S. cerevisiae), SMT3 suppressor of mif two 3 homolog 1 (yeast), Smt3C, SUMO-1, Ubiquitin-homology domain protein PIC1, ubiquitin-like 1 (sentrin), Ubiquitin-like protein SMT3C, Ubiquitin-like protein UBL1 | |
| Recombinant Human SUMO1 protein | |
| 50 μg | |
| Alzheimers Research, Core ESC Like Genes, Neuroscience, Stem Cell Markers | |
| 7341 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction