missing translation for 'onlineSavingsMsg'
Learn More

SUMO1 Antibody, DyLight 594, Novus Biologicals™

Product Code. 18405216 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18405216 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18405216 Supplier Novus Biologicals Supplier No. NBP176737DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SUMO1 Polyclonal antibody specifically detects SUMO1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen SUMO1
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias DAP1, GAP modifying protein 1, GAP-modifying protein 1, GMP1SMT3CSMT3H3OFC10UBL1PIC1, SENP2, sentrin, small ubiquitin-related modifier 1, SMT3, SMT3 homolog 3, SMT3 suppressor of mif two 3 homolog 1 (S. cerevisiae), SMT3 suppressor of mif two 3 homolog 1 (yeast), Smt3C, SUMO-1, Ubiquitin-homology domain protein PIC1, ubiquitin-like 1 (sentrin), Ubiquitin-like protein SMT3C, Ubiquitin-like protein UBL1
Host Species Rabbit
Immunogen Antibody was raised against a 14 amino acid synthetic peptide from near the amino terminus of human sumo. The immunogen is located within the first 50 amino acids of Sumo. Amino Acid Squence: EAKPSTEDLGDKKE
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Alzheimers Research, Core ESC Like Genes, Neuroscience, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 7341
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.