missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SUMO1 Antibody (5G3), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26296-100ul
This item is not returnable.
View return policy
Description
SUMO1 Monoclonal antibody specifically detects SUMO1 in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation
Specifications
| SUMO1 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation | |
| 5G3 | |
| Flow Cytometry, ELISA, Immunohistochemistry 1:50-1:200, Immunoprecipitation 1:200-1:1000 | |
| DAP1, GAP modifying protein 1, GAP-modifying protein 1, GMP1SMT3CSMT3H3OFC10UBL1PIC1, SENP2, sentrin, small ubiquitin-related modifier 1, SMT3, SMT3 homolog 3, SMT3 suppressor of mif two 3 homolog 1 (S. cerevisiae), SMT3 suppressor of mif two 3 homolog 1 (yeast), Smt3C, SUMO-1, Ubiquitin-homology domain protein PIC1, ubiquitin-like 1 (sentrin), Ubiquitin-like protein SMT3C, Ubiquitin-like protein UBL1 | |
| A synthesized peptide derived from Human SUMO1 [UniProt P63165] | |
| 100 μL | |
| Alzheimers Research, Core ESC Like Genes, Neuroscience, Stem Cell Markers | |
| 7341 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction