missing translation for 'onlineSavingsMsg'
Learn More

SUMO1 Antibody (003), Alexa Fluor™ 647, Novus Biologicals™

Product Code. 18390670 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18390670 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18390670 Supplier Novus Biologicals Supplier No. NBP290170AF647

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

SUMO1 Monoclonal specifically detects SUMO1 in Human samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SUMO1
Applications ELISA
Classification Monoclonal
Clone 3
Conjugate Alexa Fluor 647
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias DAP1, GAP modifying protein 1, GAP-modifying protein 1, GMP1SMT3CSMT3H3OFC10UBL1PIC1, SENP2, sentrin, small ubiquitin-related modifier 1, SMT3, SMT3 homolog 3, SMT3 suppressor of mif two 3 homolog 1 (S. cerevisiae), SMT3 suppressor of mif two 3 homolog 1 (yeast), Smt3C, SUMO-1, Ubiquitin-homology domain protein PIC1, ubiquitin-like 1 (sentrin), Ubiquitin-like protein SMT3C, Ubiquitin-like protein UBL1
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human SUMO1 (AAH66306.1; Ser2-Val101).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Alzheimers Research, Core ESC Like Genes, Neuroscience, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 7341
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.