missing translation for 'onlineSavingsMsg'
Learn More

SUMO-interacting Motif (SIM) Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

£328.00 - £470.00

Specifications

Antigen SUMO-interacting Motif (SIM)
Applications Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18279471
View Documents
Novus Biologicals
NBP2-55180
100 μL
£470.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18647458
View Documents
Novus Biologicals
NBP2-55180-25ul
25 μL
£328.00
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

SUMO-interacting Motif (SIM) Polyclonal specifically detects SUMO-interacting Motif (SIM) in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

SUMO-interacting Motif (SIM)
Polyclonal
Rabbit
Human
Chromosome 5 Open Reading Frame 25, Oocyte Maturation Associated 1, OOMA1, Platform Element For Inhibition Of Autolytic Degradation, PLEIAD, SIMC1, SUMO-Interacting Motif-Containing Protein 1, SUMO-Interacting Motifs Containing 1
SIMC1
IgG
Affinity Purified
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
RUO
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide
375484
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:YVIDLTRAEGENRPIATLDLTLEPVTPSQREPTSLQTCASLSGKAVMEGQVDRSSQPTARRLINSDPVDLDLVEE
Primary
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.