missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SUMO-interacting Motif (SIM) Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£328.00 - £470.00
Specifications
| Antigen | SUMO-interacting Motif (SIM) |
|---|---|
| Dilution | Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml |
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18443572
|
Novus Biologicals
NBP2-30700-25ul |
25 μL |
£328.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18133265
|
Novus Biologicals
NBP2-30700 |
0.1 mL |
£470.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SUMO-interacting Motif (SIM) Polyclonal specifically detects SUMO-interacting Motif (SIM) in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| SUMO-interacting Motif (SIM) | |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| Chromosome 5 Open Reading Frame 25, Oocyte Maturation Associated 1, OOMA1, Platform Element For Inhibition Of Autolytic Degradation, PLEIAD, SIMC1, SUMO-Interacting Motif-Containing Protein 1, SUMO-Interacting Motifs Containing 1 | |
| SIMC1 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml | |
| Polyclonal | |
| Rabbit | |
| Human | |
| Q8NDZ2 | |
| 375484 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: MKIQQLHPANAKTVEWDWKLLTYVMEEEGQTLPGRVLFLRYVVQTLEDDFQQTLRRQRQHLQQSIANMVLSCDKQPHNVRDVIKWLVKA | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title