missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SUMO Activating Enzyme E1 (SAE1) Rabbit anti-Human, Mouse, Rat, Clone: 0L2N2, Novus Biologicals™
Shop All Bio Techne ProductsDescription
SUMO Activating Enzyme E1 (SAE1) Monoclonal antibody specifically detects SUMO Activating Enzyme E1 (SAE1) in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | SUMO Activating Enzyme E1 (SAE1) |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | 0L2N2 |
| Conjugate | Unconjugated |
| Formulation | PBS, 0.05% BSA, 50% glycerol, pH7.3 |
| Gene Alias | AOS1activator of SUMO1, FLJ3091, HSPC140, Sua1, SUA1sentrin/SUMO-activating protein AOS1, SUMO-1 activating enzyme E1 N subunit, SUMO1 activating enzyme subunit 1, SUMO-1 activating enzyme subunit 1, SUMO-activating enzyme subunit 1, Ubiquitin-like 1-activating enzyme E1A, ubiquitin-like protein SUMO-1 activating enzyme, UBLE1A |
| Host Species | Rabbit |
| Immunogen | A synthetic peptide corresponding to a sequence within amino acids 200-300 of human SUMO Activating Enzyme E1 (SAE1) (Q9UBE0). |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?