missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SUMO Activating Enzyme E1 (SAE1), p Ser185 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-78069
This item is not returnable.
View return policy
Description
SUMO Activating Enzyme E1 (SAE1) Polyclonal specifically detects SUMO Activating Enzyme E1 (SAE1) in Human samples. It is validated for Western Blot, ELISA.
Specifications
| SUMO Activating Enzyme E1 (SAE1) (p Ser185) | |
| Polyclonal | |
| Unconjugated | |
| 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide | |
| AOS1activator of SUMO1, FLJ3091, HSPC140, Sua1, SUA1sentrin/SUMO-activating protein AOS1, SUMO-1 activating enzyme E1 N subunit, SUMO1 activating enzyme subunit 1, SUMO-1 activating enzyme subunit 1, SUMO-activating enzyme subunit 1, Ubiquitin-like 1-activating enzyme E1A, ubiquitin-like protein SUMO-1 activating enzyme, UBLE1A | |
| Rabbit | |
| 38.45 kDa | |
| 0.1 mg | |
| Phospho Specific, Stem Cell Markers | |
| 10055 | |
| Human, Mouse, Rat, Bovine, Canine | |
| IgG |
| Western Blot, ELISA | |
| 1.02 mg/ml | |
| Western Blot 2 ug/ml, ELISA 1:5000-1:25000 | |
| Q9UBE0 | |
| SAE1 | |
| This purified SUMO Activating Enzyme E1 (SAE1) [p Ser185] Antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to a region surrounding S185 of the human SUMO Activating Enzyme E1 (SAE1) (SAE1) protein. (Uniprot: Q9UBE0) | |
| Affinity Purified | |
| RUO | |
| Primary | |
| This purified antibody is directed against human SUMO Activating Enzyme E1 protein. This antibody is specific for human SAE1 protein phosphorylated at S185. A BLAST analysis using the sequence of the immunizing peptide was used to suggest that this antibody would react with SUMO Activating Enzyme E1 protein from human (100%), bovine, dog, chimpanzee (96%), mouse (93%), and rat (92%) based on a high degree of sequence homology. Cross reactivity against this protein from other sources has not been determined. | |
| Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction