Learn More
Invitrogen™ SUMF2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5116764
Description
Positive Samples: U-87MG, A-549, HepG2, BxPC-3 Immunogen sequence: QATSMVQLQG GRFLMGTNSP DSRDGEGPVR EATVKPFAID IFPVTNKDFR DFVREKKYRT EAEMFGWSFV FEDFVSDELR NKATQPMKSV LWWLPVEKAF WRQPAGPGSG IRERLEHPVL HVSWNDARAY CAWRGKRLPT EEEWEFAARG GLKGQVYPWG NWFQPNRTNL WQGKFPKGDK AEDGFHGVSP VNAFPAQNNY GLYDLLGNVW EWTASPYQAA EQDMRVLRGA SWIDTADGSA NHRARVTTRM GNTPDSASDN LGFRCAADAG RPPGEL.
Specifications
| SUMF2 | |
| Polyclonal | |
| Unconjugated | |
| SUMF2 | |
| 2610040F05Rik; C-alpha-formyglycine-generating enzyme 2; C-alpha-formylglycine-generating enzyme 2; DKFZp566I1024; DKFZp686I1024; DKFZp686L17160; DKFZp781L1035; FGE2; Inactive C-alpha-formylglycine-generating enzyme 2; MGC99485; Paralog of formylglycine-generating enzyme; paralog of the formylglycine-generating enzyme; pFGE; PSEC0171; sulfatase modifying factor 2; sulfatase-modifying factor 2; Sumf2; UNQ1968/PRO4500 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 25870, 360800, 67902 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 1.9 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q8BPG6, Q8NBJ7 | |
| SUMF2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 25-301 of human SUMF2 (NP_056226.3). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.