missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SUMF2 Monoclonal Antibody (330312), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA524058
This item is not returnable.
View return policy
Description
No cross-reactivity with recombinant human SUMF1 or recombinant mouse SUMF2 is observed. Reconstitute at 0.5 mg/mL in sterile PBS.
SUMF2 Monoclonal antibody specifically detects SUMF2 in Human samples. It is validated for Immunoprecipitation, Western Blot.
Specifications
| SUMF2 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| Q8NBJ7 | |
| SUMF2 | |
| Mouse myeloma cell line NS0-derived recombinant human Sulfatase Modifying Factor 2/SUMF2 Gln26-Leu301 | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Immunoprecipitation, Western Blot | |
| 330312 | |
| Unconjugated | |
| SUMF2 | |
| 2610040F05Rik; C-alpha-formyglycine-generating enzyme 2; C-alpha-formylglycine-generating enzyme 2; DKFZp566I1024; DKFZp686I1024; DKFZp686L17160; DKFZp781L1035; FGE2; Inactive C-alpha-formylglycine-generating enzyme 2; MGC99485; Paralog of formylglycine-generating enzyme; paralog of the formylglycine-generating enzyme; pFGE; PSEC0171; sulfatase modifying factor 2; sulfatase-modifying factor 2; Sumf2; UNQ1968/PRO4500 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 25870 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction