Learn More
SUMF1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 25015-1-AP-150UL
 View more versions of this product
Product Code. 16873535
  • £349.00 / 150µL
Description
This gene encodes an enzyme that catalyzes the hydrolysis of sulfate esters by oxidizing a cysteine residue in the substrate sulfatase to an active site 3-oxoalanine residue, which is also known as C-alpha-formylglycine. Mutations in this gene cause multiple sulfatase deficiency, a lysosomal storage disorder. Alternative splicing results in multiple transcript variants.
Specifications
Specifications
SUMF1
Polyclonal
Unconjugated
SUMF1
AAPA3037, FGE, sulfatase modifying factor 1, SUMF1, UNQ3037/PRO9852
Rabbit
Antigen Affinity Chromatography
RUO
285362, 58911
-20°C
Liquid
Western Blot
0.22 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q8NBK3, Q8R0F3
SUMF1
SUMF1 Fusion Protein Ag19998
150 μL
Primary
Human, Mouse