missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Sumatriptan succinate

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Sumatriptan succinate is a 5-HT1 receptor agonist (Ki values are 17, 27 and 100 nM at 5-HT1D, 5-HT1B and 5-HT1A receptors respectively). Antimigraine agent.
Specifications
Specifications
| Quantity | 10 mg |
| Product Type | Sumatriptan succinate |
| Purity | >99% |
| Molecular Weight (g/mol) | 413.49 |
| Inhibitors | Sumatriptan succinate |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction