missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Sumatriptan succinate

Product Code. 15801042 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15801042 1 g 1g
15811042 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15801042 Supplier Thermo Scientific Chemicals Supplier No. 462420010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 103628-48-4 | C18H27N3O6S | 413.49 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103628-48-4
Molecular Formula C18H27N3O6S
Molecular Weight (g/mol) 413.49
InChI Key PORMUFZNYQJOEI-UHFFFAOYSA-N
IUPAC Name 1-{3-[2-(dimethylamino)ethyl]-1H-indol-5-yl}-N-methylmethanesulfonamide; butanedioic acid
SMILES OC(=O)CCC(O)=O.CNS(=O)(=O)CC1=CC=C2NC=C(CCN(C)C)C2=C1

Specifications

Melting Point 165.0°C to 166.0°C
Color Clear
Quantity 1 g
Solubility Information Solubility aq. soln.: soluble. Other solubilities: very soluble in dimethyl sulfoxide, insoluble in ether & ethanol, slightly soluble in methanol
Formula Weight 413.49
Chemical Name or Material Sumatriptan succinate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.