missing translation for 'onlineSavingsMsg'
Learn More

Sumanirole maleate, MedChemExpress

Product Code. 16424097
Encompass_Preferred
Click to view available options
Quantity:
5 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16424097

Brand: Medchem Express HY70081A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Sumanirole maleate (U-95666E; PNU-95666E) is a highly selective D2 receptor full agonist with an ED50 of about 46 nM. Sumanirole was developed for the treatment of Parkinson's disease and restless leg syndrome.

Sumanirole maleate (U-95666E

Specifications

Chemical Name or Material Sumanirole maleate
CAS 179386-44-8
Color White
Purity Grade Notes Research
Molecular Formula C15H17N3O5
Quantity 50 mg
Synonym U-95666E PNU-95666
Solubility Information DMSO : 50 mg/mL (156.59 mM; Need ultrasonic) ∣H2O : 10 mg/mL (31.32 mM; Need ultrasonic)
SMILES O=C1NC2=C3N1C[C@H](NC)CC3=CC=C2.O=C(/C=C\C(O)=O)O
Molecular Weight (g/mol) 319.31
Formula Weight 319.31
Percent Purity 99.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.