missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SULT2B1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 29185-1-AP-150UL
This item is not returnable.
View return policy
Description
SULT2B1 Polyclonal antibody apecifically detetects SULT2B1 antigen in Human, Mouse Samples. It is validated for Western Blot, ELISA.Specifications
| SULT2B1 | |
| Polyclonal | |
| Unconjugated | |
| BC034694 | |
| Rabbit | |
| Antigen affinity purification | |
| Primary | |
| Human, Mouse | |
| Liquid |
| Western Blot, ELISA | |
| 1200 μg/mL | |
| PBS with 0.02% sodium azide and 50% glycerol pH 7.3. | |
| SULT2B1 | |
| SULT2B1 fusion protein Ag30209 | |
| 150 μL | |
| 6820 | |
| Store at -20°C. Stable for one year after shipment. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction