missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SULT1C2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5100115
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total SULT1C2.
Sulfotransferase enzymes catalyze the sulfate conjugation of many hormones, neurotransmitters, drugs, and xenobiotic compounds. These cytosolic enzymes are different in their tissue distributions and substrate specificities. The gene structure (number and length of exons) is similar among family members. SULT1C2 belongs to the SULT1 subfamily, responsible for transferring a sulfo moiety from PAPS to phenol-containing compounds. SULT1C2 belongs to a SULT subfamily that shows specificity for hydroxyarylamines. SULT1C2 catalyzes the sulfonation of p-nitrophenol and N-hydroxy-2-acetylaminofluorene, but not dopamine.
Specifications
| SULT1C2 | |
| Polyclonal | |
| Unconjugated | |
| SULT1C2 | |
| (PST)G; 1810008N17Rik; 1C member 4; HAST-I; humSULTC2; LOC100729651; MGC149521; MGC34422; mOLFST; N-hydroxyarylamine sulfotransferase; N-hydroxylarylamine sulfotransferase; phenol sulfotransferase; Phenol sulfotransferase 1c1; P-SULT; rabSULT1C2; rSULT1C2; St1c1; ST1C2; ST1C4; Stp2; sulfotransferase; sulfotransferase 1C1; Sulfotransferase 1C2; sulfotransferase 1C4; sulfotransferase family 1A phenol-preferring member 2; sulfotransferase family 1A, member 2; sulfotransferase family 1A, phenol-preferring, member 2; sulfotransferase family 1C member 2; sulfotransferase family 1C member 4; sulfotransferase family, cytosolic, 1C, member 1; sulfotransferase family, cytosolic, 1C, member 2; sulfotransferase family, cytosolic, 1C, member 3; sulfotransferase family, cytosolic, 1C, member 4; sulfotransferase family, cytosolic, 1C, member C2; sulfotransferase K1; sulfotransferase, phenol preferring 2; sulphotransferase family cytosolic 1A phenol-preferring member 2; Sult1a2; SULT1C; SULT1C#1; SULT1C#2; SULT1C1; SULT1C2; Sult1c3; SULT1C4; Sultk1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 171072, 6819, 69083 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O00338, Q9D939, Q9WUW8 | |
| SULT1C2 | |
| A synthesized peptide derived from human SULT1C2(Accession O00338), corresponding to amino acid residues M209-K259. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction