missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SULT1B1 Polyclonal Antibody
Sheep Polyclonal Antibody
Brand: Invitrogen™ PA547796
This item is not returnable.
View return policy
Description
In direct ELISAs, approximately 5% cross-reactivity with recombinant human (rh) SULT1A1 and rhSULT1A3 is observed and less than 2% cross-reactivity with rhSULT2A1, rhSULT4A1, rhSULT2B1, and rhSULT1E1 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
Sulfotransferase enzymes catalyze the sulfate conjugation of many hormones, neurotransmitters, drugs, and xenobiotic compounds. These cytosolic enzymes are different in their tissue distributions and substrate specificities. The gene structure is similar among family members. However, the total genomic length of this gene is greater than that of other SULT1 genes.
Specifications
| SULT1B1 | |
| Polyclonal | |
| Unconjugated | |
| SULT1B1 | |
| DOPA/tyrosine sulfotransferase; ST1B1; ST1B2; St2b2; sulfotransferase 1B1; sulfotransferase 1B2; sulfotransferase family 1B member 1; sulfotransferase family 1B, member 1; sulfotransferase family cytosolic 1B member 1; sulfotransferase family, cytosolic, 1B, member 1; SULT1B1; SULT1B2; Thyroid hormone sulfotransferase | |
| Sheep | |
| Antigen affinity chromatography | |
| RUO | |
| 27284 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Neutralization, Western Blot | |
| 0.2 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| O43704 | |
| SULT1B1 | |
| E. coli-derived recombinant human Cytosolic Sulfotransferase 1B1/SULT1B1 (aa 2-296). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction