missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SULT1A4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5102940
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total SULT1A3.
Sulfotransferase enzymes catalyze the sulfate conjugation of many hormones, neurotransmitters, drugs, and xenobiotic compounds. These cytosolic enzymes are different in their tissue distributions and substrate specificities. The gene structure (number and length of exons) is similar among family members. This gene encodes a phenol sulfotransferase with thermolabile enzyme activity. Four sulfotransferase genes are located on the p arm of chromosome 16, this gene and SULT1A3 arose from a segmental duplication. Read-through transcription exists between this gene and the upstream SLX1B (SLX1 structure-specific endonuclease subunit homolog B) gene that encodes a protein containing GIY-YIG domains.
Specifications
| SULT1A4 | |
| Polyclonal | |
| Unconjugated | |
| SULT1A4 | |
| aryl sulfotransferase 1A3/1A4; Catecholamine-sulfating phenol sulfotransferase; HAST3; monoamine-sulfating phenol sulfotransferase; M-PST; Placental estrogen sulfotransferase; SLX1B-SULT1A4; ST1A3; ST1A3/ST1A4; ST1A4; STM; sulfokinase; Sulfotransferase 1A3; Sulfotransferase 1A3/1A4; Sulfotransferase 1A4; sulfotransferase family 1A member 4; sulfotransferase family, cytosolic, 1A, phenol-preferring, member 4; sulfotransferase, monoamine-preferring; SULT1A3; SULT1A4; Thermolabile phenol sulfotransferase; TL-PST | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 445329 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P0DMN0 | |
| SULT1A4 | |
| A synthesized peptide derived from human SULT1A4(Accession P0DMN0), corresponding to amino acid residues G65-P115. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction