missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SULT1A3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-31809
This item is not returnable.
View return policy
Description
SULT1A3 Polyclonal specifically detects SULT1A3 in Human samples. It is validated for Western Blot.
Specifications
| Cytosolic Sulfotransferase 1A3/SULT1A3/CSPS | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal | |
| SULT1A3 | |
| Recombinant protein encompassing a sequence within the center region of human SULT1A3. The exact sequence is proprietary. | |
| Immunogen affinity purified | |
| RUO | |
| 6818 | |
| Human | |
| Purified |
| Western Blot | |
| 5.96mg/mL | |
| Western Blot 1:500-1:3000 | |
| EC 2.8.2, EC 2.8.2.1, HAST, M form) phenol sulfotransferase, MGC117469, Monoamine-sulfating phenol sulfotransferase, ST1A3/ST1A4, sulfotransferase family, cytosolic, 1A, phenol-preferring, member 3, Sulfotransferase, monoamine-preferring, Thermolabile phenol sulfotransferase | |
| Rabbit | |
| 34 kDa | |
| 0.1 mL | |
| Primary | |
| Tested in human. Predicted cross-reactivity based on sequence identity: Monkey (94%). Not yet tested in any other species. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction