missing translation for 'onlineSavingsMsg'
Learn More

SULT1A3 Antibody, DyLight 550, Novus Biologicals™

Product Code. 18342513 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18342513 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18342513 Supplier Novus Biologicals Supplier No. NBP299790R

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SULT1A3 Polyclonal specifically detects SULT1A3 in Human samples. It is validated for Western Blot, ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cytosolic Sulfotransferase 1A3/SULT1A3/CSPS
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate DyLight 550
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias EC 2.8.2, EC 2.8.2.1, HAST, M form) phenol sulfotransferase, MGC117469, Monoamine-sulfating phenol sulfotransferase, ST1A3/ST1A4, sulfotransferase family, cytosolic, 1A, phenol-preferring, member 3, Sulfotransferase, monoamine-preferring, Thermolabile phenol sulfotransferase
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Cytosolic Sulfotransferase 1A3/SULT1A3/CSPS (P50224-1; Glu2-Leu295).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6818
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.