missing translation for 'onlineSavingsMsg'
Learn More

SULT1A2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

£328.00 - £513.00

Specifications

Antigen SULT1A2
Dilution Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18424102
View Documents
Novus Biologicals
NBP2-31904-25ul
25 μL
£328.00
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18163123
View Documents
Novus Biologicals
NBP2-31904
0.1 mL
£513.00
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

SULT1A2 Polyclonal specifically detects SULT1A2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

SULT1A2
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
RUO
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Aryl sulfotransferase 2, arylamine sulfotransferase, EC 2.8.2, EC 2.8.2.1, HAST4, MGC142287, MGC142289, Phenol sulfotransferase 2, phenolic-metabolizing (P) form of PST, phenol-preferring phenol sulfotransferase2, Phenol-sulfating phenol sulfotransferase 2, P-PST 2, ST1A2, STP2P-PST, sulfotransferase 1A2, sulfotransferase family, cytosolic, 1A, phenol-preferring, member 2, thermostable phenol sulfotransferase, TSPST2
SULT1A2
IgG
Affinity Purified
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500
Polyclonal
Rabbit
Human
P50226
6799
This antibody was developed against a recombinant protein corresponding to amino acids: ELIQDISRPPLEYVKGVPLIKYFAEALGPLQSFQARPDDLL
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.